Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

50 kg
Catalog # SPCMCY110-50KGBL
Irritant
Supplier:   Spectrum Chemical
L(+)-Cysteine
Catalog # SPCMCY110-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  52-90-4
L(+)-Cysteine

50 kg

CAS Number:  52-90-4
Price
13 150,00 €
Log in for availability
1 * 50 kg
Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Boiling Pt:254 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.523 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pk:
    50 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.