Orders LinkContactUsLinkComponent
Process documentaion
APH 110 NET500 Segosoft network converter, converts serial data (RS232) to network protocol (TCP/IP), Ø 6×210 mm, G 788 (inc CD)
Catalog # MIEL9687440
Supplier:   MIELE
Process documentaion
Catalog # MIEL9687440
Supplier:  MIELE
Process documentaion
Process documentaion
APH 110 NET500 Segosoft network converter, converts serial data (RS232) to network protocol (TCP/IP), Ø 6×210 mm, G 788 (inc CD)
Price
1 163,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 110 NET500 Segosoft network converter, converts serial data (RS232) to network protocol (TCP/IP), Ø 6×210 mm
  • Seq:
    09
  • For:
    G 788 (inc CD)
  • Cat. No.:
    MIEL9687440
  • W×D×H:
    Ø 6×210 mm