Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Messpipetten 1ML:0,01, DIN-AS, KB, FUER VOLL- UND TEILABLAUF, DIN 12697, COLOR-CODE-GELB 1 * 12 items
Catalog # WITG3.510.101
Supplier:
witeg Labortechnik
Expand image
Messpipetten 1ML:0,01, DIN-AS, KB, FUER VOLL- UND TEILABLAUF, DIN 12697, COLOR-CODE-GELB 1 * 12 items
Catalog # WITG3.510.101
Supplier:
witeg Labortechnik
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Pipette stand, rotary
Supplier: BURKLE
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection