Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
200 mm, 19/26 NS, 19/26 NS
Catalog # WITG1.521.201
Supplier:   witeg Labortechnik
Fractionating columns, Vigreux
Catalog # WITG1.521.201
Supplier:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
200 mm, 19/26 NS, 19/26 NS
Price
54,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Total length:
    200 mm