Orders LinkContactUsLinkComponent
BD® Blunt Needles
BD® Blunt Needles
18 G, 40 mm, Blunt bevel, Sterile, Red
Catalog # BDAM303129UK
BD® Blunt Needles
Catalog # BDAM303129UK
Supplier:  
BD® Blunt Needles
18 G, 40 mm, Blunt bevel, Sterile, Red
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Needle Blunt Fill 18G 1 1/2inch sterile 1 * 100 items Catalog #BDAM303129
Product BDAM303129 is a replacement for BDAM303129UK but varies by Brand, Packaging or Quantity.
Product Details & Documents
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
  • The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
  • The blunt filter is designed for safe ampoule medication preparation
  • BD blunt fill needles have female luer fittings, which mate to male luer fittings
  • BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
  • Natural rubber latex is not part of the material formulation
  • Sterile, singe use
Caution: Note: The BD blunt fill and filter needles are not for skin injection.
Specifications
  • Colour code:
    Red
  • Gauge:
    18 G
  • Type:
    Sterile
  • Type of tip:
    Blunt bevel
  • Length:
    40 mm
  • Seq:
    04