ENDOTELL COLLAGEN: COLLAGEN REAGENT FOR PATELET AGGREGATION TESTS 1 * 1 ST
Catalog # MSPPHY-AG005K
Supplier: MARKET SOURCE PART PROCESS
ENDOTELL COLLAGEN: COLLAGEN REAGENT FOR PATELET AGGREGATION TESTS 1 * 1 ST
Catalog # MSPPHY-AG005K
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Laboratory marker
Supplier: MARIENFELD
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 99.5 % denatured with 1 % MEK 99,5% ≥99.5%, GPR RECTAPUR® (denatured with 1% MEK)
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals