Order Entry
Germany
Orders LinkContactUsLinkComponent
Carts

Carts

Enclosed linen exchange cart, 1080×709×1990 mm
Catalog # METOSXRD76CM3
Supplier:   Metro International
Carts
Catalog # METOSXRD76CM3
Carts
Carts

Enclosed linen exchange cart, 1080×709×1990 mm

Price
10 810,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The Starsys carts are available in a wide variety of heights and system widths and are ideal for the storage needs in the laboratories. The carts features smooth polymer top/worksurface and are provided with 127 mm polymer swivel casters, the front two casters comes with total-lock toe brake to lock both the wheel and the horn.

  • Easy to clean
  • Interchangeable drawers (76, 152 and 230 mm) with removable totes can be fully extended for easy access
  • Colour-coded drawer pulls are available in an array of choices
  • Label holders available to quickly identify drawer contents
Specifications
  • Description:
    Enclosed linen exchange cart
  • W×D×H:
    1080×709×1990 mm
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.