Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cyclohexane ACS

Plastic drum, 5 L
Catalog # ACRO446280050
Irritant
Health
Environment
Flammable
Cyclohexane ACS
Catalog # ACRO446280050
CAS Number:  110-82-7
Cyclohexane ACS
Cyclohexane ACS

Plastic drum, 5 L

CAS Number:  110-82-7
Price
123,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:81 °C (760 mmHg)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • UN:1145
  • ADR:3,II

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Substance darkened by H2SO4:
    Passes test

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.