Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Lanolin, anhydrous USP

Plastic drum, 2,5 kg
Catalog # ACRO413000025
Lanolin, anhydrous USP
Catalog # ACRO413000025
CAS Number:  8006-54-0
Lanolin, anhydrous USP
Lanolin, anhydrous USP

Plastic drum, 2,5 kg

CAS Number:  8006-54-0
Price
236,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • Merck Index:12,05371
  • Melting Pt:38…40 °C
  • Density:0,932 to 0.945 g/cm³ (15 °C)
  • Flash Pt:209 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:232-348-6
Specifications

Specification Test Results

  • Appearance:
    Yellow Wax
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 44 °C
  • Water:
    ≤0.25 %
  • Additional info:
    Water-soluble acids and alkalies: passes test
  • Antioxidant:
    ≤0.02 % (BHT)
  • Residue after ignition:
    ≤0.1 %
  • Acidity:
    ≤2.0 ml (ml 0.1 N NaOH)
  • Alkalinity:
    Passes test
  • Chloride (Cl):
    ≤0.035 %
  • Iodine value:
    18 to 36
  • Ammonia (NH3):
    Passes test

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 kg

Specifications