Order Entry
Germany
Orders LinkContactUsLinkComponent
Methyl red, pure indicator

Methyl red, pure indicator

Glass bottle, 500 g
Catalog # ACRO264445000
Methyl red, pure indicator
Catalog # ACRO264445000
CAS Number:  493-52-7
Methyl red, pure indicator
Methyl red, pure indicator

Glass bottle, 500 g

CAS Number:  493-52-7
Price
383,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1,31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark red to brown to purple to violet Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    178 to 182 °C
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥650 (430 nm, in buffer)
  • Visual transition interval:
    from pH 4.2 (pink or red) to pH 6.2 (yellow)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications