Sodium butyrate 98%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Butyric acid sodium salt, Sodium Butyrate
- CAS Number:156-54-7
- Formula:C₄H₇NaO₂
- MDL Number:MFCD00002816
- Molecular Weight:110.09 g/mol
- Melting Pt:250…253 °C
- Density:1,324 g/cm³ (30 °C)
- Storage Temperature:Ambient
- EINECS:205-857-6
Documents
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration with HClO4:97.5 to 102.5 %
Specifications
- Pack type:Glass bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



