Order Entry
Germany
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Glass bottle, 10 mg
Catalog # ACRO337050100
Irritant
D(-)-Luciferin
Catalog # ACRO337050100
CAS Number:  2591-17-5
D(-)-Luciferin
D(-)-Luciferin

Glass bottle, 10 mg

CAS Number:  2591-17-5
Price
193,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • Melting Pt:200 °C
  • Storage Temperature:Freezer
  • EINECS:219-981-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Off-white to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2.0 % (K.F.)
  • HPLC:
    ≥97.5 %
  • UV Lambda max:
    327 to 331 nm (c=1, CH3OH)
  • Specific optical rotation:
    -32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
  • Molar extinction coefficient:
    ≥18200 l/(mol.cm) (327 to 331 nm, c=1, CH3OH)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg

Specifications