Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyridine 99+% for analysis

Pyridine 99+% for analysis

Glass bottle, 100 mL
Catalog # ACRO290861000
Irritant
Flammable
Pyridine 99+% for analysis
Catalog # ACRO290861000
CAS Number:  110-86-1
Pyridine 99+% for analysis
Pyridine 99+% for analysis

Glass bottle, 100 mL

CAS Number:  110-86-1
Price
31,60 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Boiling Pt:115…116 °C (760 mmHg)
  • Melting Pt:–42 °C
  • Density:0.978 g/cm³
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • UN:1282
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Non-volatile matter:
    ≤0.002 %
  • Ammonia (NH3):
    ≤0.002 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.