Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 80

Polysorbate 80

Plastic drum, 10 L
Catalog # ACRO278630100
Polysorbate 80
Catalog # ACRO278630100
CAS Number:  9005-65-6
Polysorbate 80
Polysorbate 80

Plastic drum, 10 L

CAS Number:  9005-65-6
Price
628,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:-21 °C
  • Density:1.08 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to amber Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4700 to 1.4760 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    65 to 80 mg KOH/g
  • Saponification value:
    45 to 55 mg KOH/g

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.