(S)-(+)-2-Hydroxyisovaleric acid 99+%
Glass bottle, 5 g
: (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
- :17407-55-5
- :C₅H₁₀O₃
- :MFCD00066443
- :118.13 g/mol
- :65…68 °C
- :Refrigerator
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Melting point:65 to 70 °C
- GC:≥99.0 %
- Specific optical rotation:+15° to +19° (24 °C, 589 nm) (c=2, CHCl3)
- Titration with NaOH:≥99.0 %
- Pack type:Glass bottle
- Pk:5 g



