Triethoxysilane 95%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:998-30-1
- Formula:C₆H₁₆O₃Si
- MDL Number:MFCD00009061
- Molecular Weight:164.28 g/mol
- Boiling Pt:134…135 °C (760 mmHg)
- Melting Pt:–170 °C
- Density:0.89 g/cm³
- Flash Pt:26 °C
- Storage Temperature:Ambient
- UN:3489
- ADR:6.1,I
Documents
ACRO16481
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3770 to 1.3790 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



