Triethoxysilane 95%
Glass bottle, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:998-30-1
- Formula:C₆H₁₆O₃Si
- MDL Number:MFCD00009061
- Molecular Weight:164.28 g/mol
- Boiling Pt:134…135 °C (760 mmHg)
- Melting Pt:–170 °C
- Density:0.89 g/cm³
- Flash Pt:26 °C
- Storage Temperature:Ambient
- UN:3489
- ADR:6.1,I
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3770 to 1.3790 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:10 g



