Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 98.5%, pellets for analysis

Sodium hydroxide 98.5%, pellets for analysis

Fibre drum, 25 kg
Catalog # ACRO206060250
Corrosive
Sodium hydroxide 98.5%, pellets for analysis
Catalog # ACRO206060250
CAS Number:  1310-73-2
Sodium hydroxide 98.5%, pellets for analysis
Sodium hydroxide 98.5%, pellets for analysis

Fibre drum, 25 kg

CAS Number:  1310-73-2
Price
386,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C (760 mmHg)
  • Melting Pt:318 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White Pellets
  • Titration with HCl:
    ≥98.0 %
  • Chloride (Cl):
    ≤70 ppm
  • Phosphate (PO4):
    ≤3 ppm
  • Sulfate (SO4):
    ≤40 ppm
  • Sodium carbonate (Na2CO3):
    ≤0.5 %
  • Aluminium (Al):
    ≤5 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Iron (Fe):
    ≤8 ppm
  • Nickel (Ni):
    ≤3 ppm
  • Potassium (K):
    ≤0.1 %
  • Silver (Ag):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Silicon dioxide (SiO2):
    ≤20 ppm
  • Nitrogen compounds (as N):
    ≤0.0003 %

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.