Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 98.5%, pellets for analysis

Sodium hydroxide 98.5%, pellets for analysis

Plastic drum, 10 kg
Catalog # ACRO206060100
Corrosive
Sodium hydroxide 98.5%, pellets for analysis
Catalog # ACRO206060100
CAS Number:  1310-73-2
Sodium hydroxide 98.5%, pellets for analysis
Sodium hydroxide 98.5%, pellets for analysis

Plastic drum, 10 kg

CAS Number:  1310-73-2
Price
186,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C (760 mmHg)
  • Melting Pt:318 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White Pellets
  • Titration with HCl:
    ≥98.0 %
  • Chloride (Cl):
    ≤70 ppm
  • Phosphate (PO4):
    ≤3 ppm
  • Sulfate (SO4):
    ≤40 ppm
  • Sodium carbonate (Na2CO3):
    ≤0.5 %
  • Aluminium (Al):
    ≤5 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Iron (Fe):
    ≤8 ppm
  • Nickel (Ni):
    ≤3 ppm
  • Potassium (K):
    ≤0.1 %
  • Silver (Ag):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Silicon dioxide (SiO2):
    ≤20 ppm
  • Nitrogen compounds (as N):
    ≤0.0003 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications