Sodium nitrate 99+% for analysis
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Sodium nitrate 99+% for analysis
Catalog # ACRO205962500
Supplier: Thermo Fisher Scientific
CAS Number: 7631-99-4
Jump to:
Product Details & Documents
- CAS Number:7631-99-4
- Formula:NaNO₃
- MDL Number:MFCD00011119
- Molecular Weight:84.99 g/mol
- Merck Index:13,08720
- Boiling Pt:380 °C (1013 hPa)
- Melting Pt:306 °C
- Density:2,26 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-554-3
- UN:1498
- ADR:5.1,III
- REACH:01-2119488221-41
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- pH:5.5 to 8.3 (5 % at 25°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤10 ppm
- Nitrite (NO2):≤10 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤30 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤3 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



