Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium nitrate 99+% for analysis

Potassium nitrate 99+% for analysis

Plastic drum, 10 kg
Catalog # ACRO205910100
Oxidizing
Potassium nitrate 99+% for analysis
Catalog # ACRO205910100
CAS Number:  7757-79-1
Potassium nitrate 99+% for analysis
Potassium nitrate 99+% for analysis

Plastic drum, 10 kg

CAS Number:  7757-79-1
Price
358,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Boiling Pt:400 °C (760 mmHg)
  • Melting Pt:334 °C
  • Density:2,109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystals
  • pH:
    5.5 to 8 (5 % at 20°C)
  • Titration after Ion exchange:
    ≥99.0 %
  • Chloride (Cl):
    ≤20 ppm
  • Iodate (IO3):
    ≤5 ppm
  • Nitrite (NO2):
    ≤10 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Sodium (Na):
    ≤200 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.