Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium nitrate 99+% for analysis

Potassium nitrate 99+% for analysis

Plastic bottle, 2,5 kg
Catalog # ACRO205910025
Oxidizing
Potassium nitrate 99+% for analysis
Catalog # ACRO205910025
CAS Number:  7757-79-1
Potassium nitrate 99+% for analysis
Potassium nitrate 99+% for analysis

Plastic bottle, 2,5 kg

CAS Number:  7757-79-1
Price
126,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Boiling Pt:400 °C (760 mmHg)
  • Melting Pt:334 °C
  • Density:2,109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystals
  • pH:
    5.5 to 8 (5 % at 20°C)
  • Titration after Ion exchange:
    ≥99.0 %
  • Chloride (Cl):
    ≤20 ppm
  • Iodate (IO3):
    ≤5 ppm
  • Nitrite (NO2):
    ≤10 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Sodium (Na):
    ≤200 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications