Sodium nitrite 98.5% for analysis
Plastic bottle, 5 kg
Supplier: Thermo Fisher Scientific
Sodium nitrite 98.5% for analysis
Catalog # ACRO217600050
Supplier: Thermo Fisher Scientific
CAS Number: 7632-00-0
Jump to:
Product Details & Documents
Synonyms: Diazotizing salts
- CAS Number:7632-00-0
- Formula:NaNO₂
- MDL Number:MFCD00011118
- Molecular Weight:69 g/mol
- Merck Index:13,08721
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:271 °C
- Density:2,168 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-555-9
- UN:1500
- ADR:5.1,III
Documents
ACRO21760
SDS
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder or crystals
- Titration with KMnO4:≥98.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤50 ppm
- Calcium (Ca):≤25 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.003 % (in water)
Specifications
- Pack type:Plastic bottle
- Pk:5 kg



