(S)-(-)-1,1'-Bi-2-naphthol 99%
Glass bottle, 25 g
: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
- :18531-99-2
- :C₂₀H₁₄O₂
- :MFCD00004068
- :286.33 g/mol
- :205…211 °C
- :Ambient
- :6.1,III
- Appearance:White to light grey Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:205 to 211 °C
- HPLC:≥98.5 %
- Specific optical rotation:-33° to -38° (20 °C, 589 nm) (c=1, THF)
- Pack type:Glass bottle
- Pk:25 g



