4-Chlorophenol 99+%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: p-Chlorophenol
- CAS Number:106-48-9
- Formula:C₆H₅ClO
- MDL Number:MFCD00002318
- Molecular Weight:128.56 g/mol
- Merck Index:13,02173
- Boiling Pt:220 °C (760 mmHg)
- Melting Pt:41…45 °C
- Density:1.26 g/cm³
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:203-402-6
- UN:2020
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to light brown Crystals or fused mass
- Infrared spectrum:Conforms
- GC:≥99.0 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



