α-Cyclodextrine hydrate 98+%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
α-Cyclodextrine hydrate 98+%
Catalog # ACRO227291000
Supplier: Thermo Fisher Scientific
CAS Number: 51211-51-9
Jump to:
Product Details & Documents
Synonyms: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
- CAS Number:51211-51-9
- Formula:C₃₆H₆₀O₃₀·xH₂O
- MDL Number:MFCD00150804
- Molecular Weight:972.86 g/mol (anhy)
- Melting Pt:278 °C (dec.)
- Storage Temperature:Ambient
- EINECS:233-007-4
Documents
ACRO22729
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤11 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.0 % (ELSD) on dry substance
- Specific optical rotation:+145° to +151° (20°C, 589 nm) (c=2, water) (on dry substance)
- Heavy metals (as Pb):≤15 ppm
Specifications
- Pack type:Glass bottle
- Pk:100 g



