Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Pyrene 98%

Glass bottle, 500 g
Catalog # ACRO180835000
Environment
Pyrene 98%
Catalog # ACRO180835000
CAS Number:  129-00-0
Pyrene 98%
Pyrene 98%

Glass bottle, 500 g

CAS Number:  129-00-0
Price
312,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:129-00-0
  • Formula:C₁₆H₁₀
  • MDL Number:MFCD00004136
  • Molecular Weight:202.26 g/mol
  • Merck Index:12,08147
  • Boiling Pt:393 °C (760 mmHg)
  • Melting Pt:148…152 °C
  • Density:1,27 g/cm³ (20 °C)
  • Flash Pt:210 °C
  • Storage Temperature:Ambient
  • EINECS:204-927-3
  • UN:3077
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Light yellow to yellow to yellow-green Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    147 to 152 °C
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications