Diethyl ether 99+% stabilised, extra pure
Metal drum, 25 L
Hersteller: Thermo Fisher Scientific
Diethyl ether 99+% stabilised, extra pure
Artikelnummer ACRO176830250
Hersteller: Thermo Fisher Scientific
CAS-Nummer: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Documents
ACRO17683
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 7 ppm BHT
- Relative density:(20°C) 0.714 to 0.716
- Residue after evaporation:≤0.005 %
- Acidity:≤0.001 meq/g
- Peroxides:≤0.0005 %
Specifications
- Pack type:Metal drum
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



