Order Entry
Germany
Orders LinkContactUsLinkComponent
Fluorescamine, pure

Fluorescamine, pure

Glass bottle, 2,5 g
Catalog # ACRO191670025
Irritant
Fluorescamine, pure
Catalog # ACRO191670025
CAS Number:  38183-12-9
Fluorescamine, pure
Fluorescamine, pure

Glass bottle, 2,5 g

CAS Number:  38183-12-9
Price
1 740,00 €
Log in for availability
1 * 2,5 g
Product Details & Documents

CAS No.: 38183-12-9

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Melting Pt:155…157 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    155 to 157 °C
  • Specific extinction E (1%/1cm):
    ≥900 (235 nm, ether)
  • Suitability:
    fluorescence ind. for prim. amines and amino acids

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.