n-Eicosane 99%
Glass bottle, 500 g
: N-Eicosane
- :112-95-8
- :H₃C(CH₂)₁₈CH₃
- :MFCD00009344
- :282.55 g/mol
- :220 °C (30 mmHg)
- :36…38 °C
- :0.7886 g/cm³
- :>112 °C
- :Ambient
- :204-018-1
- Appearance:Transparent white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:36 to 38 °C
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:500 g



