Order Entry
Germany
Orders LinkContactUsLinkComponent
Anthrone ACS

Anthrone ACS

Plastic bottle, 100 g
Catalog # ACRO104961000
Irritant
Anthrone ACS
Catalog # ACRO104961000
CAS Number:  90-44-8
Anthrone ACS
Anthrone ACS

Plastic bottle, 100 g

CAS Number:  90-44-8
Price
431,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Boiling Pt:721 °C (760 mmHg)
  • Melting Pt:152…157 °C
  • Storage Temperature:Ambient
  • EINECS:201-994-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    152 to 157 °C
  • Solubility:
    (in ethyl acetate) Passes test
  • Absorbance:
    ≤0.75 (425 nm, 1 cm cell)
  • Sensitivity:
    (to carbohydrates) Passes test

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.