Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Methylmorpholine 99%

4-Methylmorpholine 99%

Plastic drum, 2,5 kg
Catalog # ACRO127150025
Irritant
Corrosive
Flammable
4-Methylmorpholine 99%
Catalog # ACRO127150025
CAS Number:  109-02-4
4-Methylmorpholine 99%
4-Methylmorpholine 99%

Plastic drum, 2,5 kg

CAS Number:  109-02-4
Price
237,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115…116 °C (750 mmHg)
  • Melting Pt:–65 °C
  • Density:0.92 g/cm³
  • Flash Pt:14 °C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4339 to 1.4359 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤50 APHA

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.