Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrocatechol 99+% | Avantor

Pyrocatechol 99+%

Fibre drum, 10 kg
Catalog # ACRO158980100
Corrosive
Toxic
Health
Pyrocatechol 99+%
Catalog # ACRO158980100
CAS Number:  120-80-9
Pyrocatechol 99+%
Pyrocatechol 99+%

Fibre drum, 10 kg

CAS Number:  120-80-9
Price
703,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (760 mmHg)
  • Melting Pt:103…106 °C
  • Density:1,0548 g/cm³ (20 °C)
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Off-white to grey-brown Crystalline flakes (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    103 to 106 °C
  • GC:
    ≥99.0 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.05 % (in water)

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.