Order Entry
Germany
Orders LinkContactUsLinkComponent
Xanthine 98%

Xanthine 98%

Glass bottle, 100 g
Catalog # ACRO149171000
Irritant
Xanthine 98%
Catalog # ACRO149171000
CAS Number:  69-89-6
Xanthine 98%
Xanthine 98%

Glass bottle, 100 g

CAS Number:  69-89-6
Price
442,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
  • CAS Number:69-89-6
  • Formula:C₅H₄N₄O₂
  • MDL Number:MFCD00078453
  • Molecular Weight:152.11 g/mol
  • Merck Index:12,10193
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:200-718-6

Dokumente

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C, 5 hrs)
  • Sulfated ash:
    ≤0.2 % (800°C)
  • HPLC:
    ≥97.5 %
  • TLC:
    Homogeneous
  • UV spectrophotometry:
    98 to 102 % (lambda max. 278 nm, pH 10)
  • Appearance of solution:
    (c=1, 1M NaOH) clear colorless to light yellow soln
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.