2,4,6-Trimethylaniline 97%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
2,4,6-Trimethylaniline 97%
Catalog # ACRO157965000
Supplier: Thermo Fisher Scientific
CAS Number: 88-05-1
Jump to:
Product Details & Documents
- CAS Number:88-05-1
- Formula:C₉H₁₃N
- MDL Number:MFCD00007740
- Molecular Weight:135.21 g/mol
- Boiling Pt:233 °C (760 mmHg)
- Melting Pt:–5 °C
- Density:0.96 g/cm³
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:201-794-3
- UN:2810
- ADR:6.1,III
Documents
ACRO15796
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5505 to 1.5525 (20°C, 589 nm)
- GC:≥96.0 %
- Appearance of solution:Clear colorless
- Residue after evaporation:≤0.05 %
- Related substances:≤0.15 % (Benzaldehyde)
- Acidity:Passes test
- Peroxide value:≤5
Specifications
- Pack type:Glass bottle
- Pk:500 g



