2,4,6-Trimethylaniline 97%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
2,4,6-Trimethylaniline 97%
Catalog # ACRO157960025
Supplier: Thermo Fisher Scientific
CAS Number: 88-05-1
Jump to:
Product Details & Documents
- CAS Number:88-05-1
- Formula:C₉H₁₃N
- MDL Number:MFCD00007740
- Molecular Weight:135.21 g/mol
- Boiling Pt:233 °C (760 mmHg)
- Melting Pt:–5 °C
- Density:0.96 g/cm³
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:201-794-3
- UN:2810
- ADR:6.1,III
Documents
ACRO15796
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5505 to 1.5525 (20°C, 589 nm)
- GC:≥96.0 %
- Appearance of solution:Clear colorless
- Residue after evaporation:≤0.05 %
- Related substances:≤0.15 % (Benzaldehyde)
- Acidity:Passes test
- Peroxide value:≤5
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



