Catechol violet, pure indicator grade
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Catechol violet, pure indicator grade
Catalog # ACRO146540250
Supplier: Thermo Fisher Scientific
CAS Number: 115-41-3
Jump to:
Product Details & Documents
Synonyms: Pyrocatechol Violet
- CAS Number:115-41-3
- Formula:C₁₉H₁₄O₇S
- MDL Number:MFCD00005868
- Molecular Weight:386.38 g/mol
- Storage Temperature:Ambient
- EINECS:204-088-3
Documents
Specifications
Specification Test Results
- Appearance:Green to dark green to brown Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Solubility:(0.1 % in water) Clear
- UV Lambda max:441 to 449 nm (in methanol)
- Suitability:(as complexom. indicator) Passes test
Specifications
- Pack type:Glass bottle
- Pk:25 g



