Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Uridine 99%

Glass bottle, 500 g
Catalog # ACRO140775000
Uridine 99%
Catalog # ACRO140775000
CAS Number:  58-96-8
Uridine 99%
Uridine 99%

Glass bottle, 500 g

CAS Number:  58-96-8
Price
1 830,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:162…170 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-407-5

 

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 170 °C
  • Loss on drying:
    ≤1 % (105 °C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥98.5 %
  • UV spectrophotometry:
    ≥98 % (lambda max. 262 nm, pH 7)
  • UV transmittance:
    ≥95 % (c=1, H2O, 430 nm) 1 cm cell
  • Ratio of Absorbances:
    0.72 to 0.76 (A250/A260)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications