ω,4-Dibromoacetophenone 98%
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
ω,4-Dibromoacetophenone 98%
Catalog # ACRO112682500
Supplier: Thermo Fisher Scientific
CAS Number: 99-73-0
Jump to:
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:1415 °C (760 mmHg)
- Melting Pt:107…111 °C
- Storage Temperature:Ambient
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Documents
ACRO11268
SDS
Specifications
Specification Test Results
- Appearance:Light yellow to beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:107 to 111 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



