Cyclooctanone 98%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:502-49-8
- Formula:C₈H₁₄O
- MDL Number:MFCD00001754
- Molecular Weight:126.2 g/mol
- Boiling Pt:195…197 °C (760 mmHg)
- Melting Pt:39…44 °C
- Density:0.95 g/cm³
- Flash Pt:74 °C
- Storage Temperature:Ambient
- EINECS:207-940-2
- UN:1759
- ADR:8,III
Documents
ACRO11144
SDS
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with TBAH:≥97.5 %
- Color scale:≤50 APHA
Specifications
- Pack type:Glass bottle
- Pk:25 g



