ContactUsLinkComponent
Washing Buffer PBS ********************************************** Composition:PBS-based buffer with Tween and 350 mM NaCl 10x stock solution pH-value:,,7.2 ± 0.2 Preservative:,,contains 0.045% ProClin® 300 (v/v) Storage: 10x stock solution: shelf lif 1 * 500 mL
Catalog # CAND140500
Supplier:  CANDOR BIOSCIENCE
Washing Buffer PBS ********************************************** Composition:PBS-based buffer with Tween and 350 mM NaCl 10x stock solution pH-value:,,7.2 ± 0.2 Preservative:,,contains 0.045% ProClin® 300 (v/v) Storage: 10x stock solution: shelf lif 1 * 500 mL
Catalog # CAND140500
Supplier:  CANDOR BIOSCIENCE