Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Carts

Cold room/buffer cart, 1080×632×1060 mm
Catalog # METOSXRD43CM3
Supplier:   Metro International
Carts
Catalog # METOSXRD43CM3
Carts
Carts

Cold room/buffer cart, 1080×632×1060 mm

Price
9 760,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The Starsys carts are available in a wide variety of heights and system widths and are ideal for the storage needs in the laboratories. The carts features smooth polymer top/worksurface and are provided with 127 mm polymer swivel casters, the front two casters comes with total-lock toe brake to lock both the wheel and the horn.

  • Easy to clean
  • Interchangeable drawers (76, 152 and 230 mm) with removable totes can be fully extended for easy access
  • Colour-coded drawer pulls are available in an array of choices
  • Label holders available to quickly identify drawer contents
Specifications
  • Description:
    Cold room/buffer cart
  • W×D×H:
    1080×632×1060 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.