ContactUsLinkComponent
BN-13/100, Adapter set for R-12/10 - Set of 12 adapters - made of POM-C - max. temperature 100°C - for vacutainers 4-8mL - dimensions (Ø x length): 13x100mm 1 * 1 items
Catalog # BSANBS-010208-QK
Supplier:  BIOSAN
BN-13/100, Adapter set for R-12/10 - Set of 12 adapters - made of POM-C - max. temperature 100°C - for vacutainers 4-8mL - dimensions (Ø x length): 13x100mm 1 * 1 items
Catalog # BSANBS-010208-QK
Supplier:  BIOSAN
Frequently Bought Together