ContactUsLinkComponent
R-12/10, Swing-out rotor for round bottom centrifuge tube - Dimensions (Ø × length): 16 × 105 mm - Capacity: 12 - Tube’s volume: 10–15 ml - Max. speed: 4,200 rpm - Max. RCF: LMC-3000 --> 1,700 × g - Tube Manufacturers: Nunc, Greiner, Greiner Bio-one, 1 * 1 items
Catalog # BSANBS-010208-BK
Supplier:  BIOSAN
R-12/10, Swing-out rotor for round bottom centrifuge tube - Dimensions (Ø × length): 16 × 105 mm - Capacity: 12 - Tube’s volume: 10–15 ml - Max. speed: 4,200 rpm - Max. RCF: LMC-3000 --> 1,700 × g - Tube Manufacturers: Nunc, Greiner, Greiner Bio-one, 1 * 1 items
Catalog # BSANBS-010208-BK
Supplier:  BIOSAN
Frequently Bought Together