Order Entry
Germany
Orders LinkContactUsLinkComponent
FORMALDEHYDE 35 % KLINIPATH

FORMALDEHYDE 35 % KLINIPATH

Catalog # VWRK4074-9200
Supplier:   KLINIPATH
FORMALDEHYDE 35 % KLINIPATH
Catalog # VWRK4074-9200
Supplier:  KLINIPATH
CAS Number:  
FORMALDEHYDE 35 % KLINIPATH
CAS Number:  
Price
769,00 €
Log in for availability
1 * 220 kg
Product Details & Documents