Bucket Liners
Disposable bucket liners, double bagged, irradiated, LDPE/nylon, 609,6×609,6 mm
Catalog # MICRBKL-2424IR
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.
- Laminate is 75% LDPE, 25% nylon
- Excellent for disinfectants and cleaning solutions
- Ideal for product parts transport
Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.
: Individually double packed, packed in a case of 24/50/200.
Documents
Specifications
- Description:Disposable bucket liners, double bagged, irradiated
- L×W:609,6×609,6 mm
- Material:LDPE/nylon
- Seq:02



