Orders LinkContactUsLinkComponent
Squeegee mop and handles
Squeegee mop and handles
Cleanroom handle, length 1,45 m
Catalog # PPSA3590200
Squeegee mop and handles
Catalog # PPSA3590200
Squeegee mop and handles
Squeegee mop and handles
Cleanroom handle, length 1,45 m
Price
60,97 €
Log in for availability
Product Details & Documents
Squeegee mops and handles for hygienically sensitive areas.
  • Mop: PP (body), TPE (lip)
  • Hygienic squeegee is suitable with cleanroom and telescopical handles
  • Handles are anodised aluminum tubes
Specifications
  • Description:
    Cleanroom handle, length 1,45 m
  • Seq:
    02