Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLUBILITY / STABILITY SCREEN 2 1 * 0,5 ml
Catalog # HMPTHR2-413
Supplier:
HAMPTON RESEARCH
Expand image
SOLUBILITY / STABILITY SCREEN 2 1 * 0,5 ml
Catalog # HMPTHR2-413
Supplier:
HAMPTON RESEARCH