Order Entry
Germany
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

pH buffer standard solution with certificate of analysis, 4×250 ml, 10,00, 1 L
Catalog # HACHLZW9472.99
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9472.99
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 4×250 ml, 10,00, 1 L

CAS Number:  
Price
60,20 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 L
  • Description:
    pH buffer standard solution with certificate of analysis, 4×250 ml
  • pH value:
    10,00

Specifications