Order Entry
Germany
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

pH buffer standard solution with certificate of analysis, 4×250 ml, 4,01, 1 L
Catalog # HACHLZW9466.99
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9466.99
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 4×250 ml, 4,01, 1 L

CAS Number:  
Price
60,20 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 L
  • Description:
    pH buffer standard solution with certificate of analysis, 4×250 ml
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.